Learn More
Medchemexpress LLC Bomedemstat dihydrochloride | 99.6% | 592.54 | 10 MG
SDP
Catalog No. 5000344580
Encompass_Preferred
Bomedemstat (IMG-7289) dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which then alters gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Alters gene expression.
  • Shows anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.
Catalog No. 50-003-44580 Supplier Medchemexpress LLC Supplier No. HY109169C10MM/1ML
Bomedemstat (IMG-7289) dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which then alters gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Alters gene expression.
  • Shows anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.