Learn More
Medchemexpress LLC Bomedemstat dihydrochloride | 99.6% | 592.54 | 100 MG
SDP
Catalog No. 5000345293
Encompass_Preferred
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, altering gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Increases H3K4 and H3K9 methylation
  • Alters gene expression
  • Inhibits cancer cell proliferation
  • Induces apoptosis
Catalog No. 50-003-45293 Supplier Medchemexpress LLC Supplier No. HY109169C10MG
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, altering gene expression. This compound exhibits anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Increases H3K4 and H3K9 methylation
  • Alters gene expression
  • Inhibits cancer cell proliferation
  • Induces apoptosis