Learn More
Medchemexpress LLC Bomedemstat dihydrochloride | 99.6% | 592.54 | 50 MG
SDP
Catalog No. 5000344512
Encompass_Preferred
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation to alter gene expression, showing anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active.
  • Irreversible LSD1 inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Alters gene expression.
  • Exhibits anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.
  • Selectively inhibits proliferation and induces apoptosis of Jak2V617F cells.
  • Enhances cell survival and induces apoptosis via BCL-XL and PUMA.
  • Leads to cell cycle arrest.
  • Normalizes or improves blood cell counts in animal models.
  • Reduces spleen volumes in animal models.
  • Restores normal splenic architecture.
  • Reduces bone marrow fibrosis in animal models.
Catalog No. 50-003-44512 Supplier Medchemexpress LLC Supplier No. HY109169C50MG
Bomedemstat dihydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation to alter gene expression, showing anti-cancer activities by inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active.
  • Irreversible LSD1 inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Alters gene expression.
  • Exhibits anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.
  • Selectively inhibits proliferation and induces apoptosis of Jak2V617F cells.
  • Enhances cell survival and induces apoptosis via BCL-XL and PUMA.
  • Leads to cell cycle arrest.
  • Normalizes or improves blood cell counts in animal models.
  • Reduces spleen volumes in animal models.
  • Restores normal splenic architecture.
  • Reduces bone marrow fibrosis in animal models.