Learn More
Medchemexpress LLC Bomedemstat dihydrochloride | 99.6% | 592.54 g/mol | C28H36Cl2FN7O2 | 5 MG
SDP
Catalog No. 5000264077
Encompass_Preferred
Bomedemstat dihydrochloride is the dihydrochloride salt of bomedemstat, an irreversible lysine-specific demethylase 1 (LSD1) inhibitor supplied for research use. It is provided as a white to light yellow solid intended for biochemical and cell-based studies of epigenetic regulation and oncology-related research. Store sealed, away from moisture at -20°C; stability in solution: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-64077 Supplier Medchemexpress LLC Supplier No. HY109169C5MG
Bomedemstat dihydrochloride is the dihydrochloride salt of bomedemstat, an irreversible lysine-specific demethylase 1 (LSD1) inhibitor supplied for research use. It is provided as a white to light yellow solid intended for biochemical and cell-based studies of epigenetic regulation and oncology-related research. Store sealed, away from moisture at -20°C; stability in solution: -80°C up to 6 months, -20°C up to 1 month.