Learn More
Medchemexpress LLC Bomedemstat hydrochloride | 99.9% | 556.07 g/mol | C28H35ClFN7O2 | 5 MG
SDP
Catalog No. 5000264630
Encompass_Preferred
Bomedemstat hydrochloride is the hydrochloride salt of bomedemstat, an orally active, irreversible lysine-specific demethylase 1 (LSD1) inhibitor used in preclinical research to modulate histone methylation and evaluate antineoplastic activity. Supplied as a high-purity solid for research use.
  • Irreversible LSD1 inhibitor.
  • Hydrochloride salt for improved solubility.
  • High purity (≈99.9%).
  • Modulates H3K4 and H3K9 methylation.
  • Suitable for in vitro and preclinical studies.
  • Available as solid and as a 10 mM DMSO solution.
Catalog No. 50-002-64630 Supplier Medchemexpress LLC Supplier No. HY109169B5MG
Bomedemstat hydrochloride is the hydrochloride salt of bomedemstat, an orally active, irreversible lysine-specific demethylase 1 (LSD1) inhibitor used in preclinical research to modulate histone methylation and evaluate antineoplastic activity. Supplied as a high-purity solid for research use.
  • Irreversible LSD1 inhibitor.
  • Hydrochloride salt for improved solubility.
  • High purity (≈99.9%).
  • Modulates H3K4 and H3K9 methylation.
  • Suitable for in vitro and preclinical studies.
  • Available as solid and as a 10 mM DMSO solution.