Learn More
Medchemexpress LLC Bomedemstat hydrochloride | 99.94% | 556.07 | 10 MILLIGRAM
SDP
Catalog No. 5000344671
Encompass_Preferred
Bomedemstat hydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which alters gene expression. This compound exhibits anti-cancer activities, inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Shows anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.
Catalog No. 50-003-44671 Supplier Medchemexpress LLC Supplier No. HY109169B10MG
Bomedemstat hydrochloride is an orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor. It increases H3K4 and H3K9 methylation, which alters gene expression. This compound exhibits anti-cancer activities, inhibiting cancer cell proliferation and inducing apoptosis.
  • Orally active and irreversible lysine-specific demethylase 1 (LSD1) inhibitor.
  • Increases H3K4 and H3K9 methylation.
  • Shows anti-cancer activities.
  • Inhibits cancer cell proliferation.
  • Induces apoptosis.