Learn More
SPEX CERTIPREP LLC 3P BOMIDE 500ML 1000PPM
Supplier:  SPEX CERTIPREP LLC 3P ICBRM500
Encompass_Preferred
(HPS by Zeptometrix) Bromide (from NaBr) IC Standard 10.00mg/mL in H2O Exp. date: 18 Months
Catalog No. NC9916338
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.