Learn More
Being Scientific BON-30T - 1.1 cu ft (30 Liters)Temperature Range:Amb+10°C - 300°C .
Catalog No. 5000217542
Encompass_Preferred
The BON Natural Convection Drying Oven series offers low air turbulences for gentle drying and heating. This is ideal for sterilization, powder drying and high temperature storage. The 4.3-inch touchscreen controller provides fast, accurate, and uniform heat displacement. Users can preset 8 periods and 8 steps for a total of 64 programs. Temperature and time can be set and programmed at the same time. The doors are engineered with airtight sealing and inner chambers are designed with corrosion resistant stainless steel. Safety alarms are installed for – overtemperature, over current protection, and temperature deviation. These ovens provide excellent temperature stability and reproducibility for all your desired application needs ensuring maximum efficiency, safety, and ease of use.
Catalog No. 50-002-17542 Supplier Being Scientific Supplier No. BO211030U
The BON Natural Convection Drying Oven series offers low air turbulences for gentle drying and heating. This is ideal for sterilization, powder drying and high temperature storage. The 4.3-inch touchscreen controller provides fast, accurate, and uniform heat displacement. Users can preset 8 periods and 8 steps for a total of 64 programs. Temperature and time can be set and programmed at the same time. The doors are engineered with airtight sealing and inner chambers are designed with corrosion resistant stainless steel. Safety alarms are installed for – overtemperature, over current protection, and temperature deviation. These ovens provide excellent temperature stability and reproducibility for all your desired application needs ensuring maximum efficiency, safety, and ease of use.