Learn More
Leica Biosystems Bond Aspirating Probe
Supplier:  Leica Biosystems S21.0605
Encompass
The BOND Aspirating Probe Cleaning System contains reagents optimized to clean the aspirating probe of residual DAB.Sold in a standard reagent tray, the system is loaded onto BOND where a predefined cleaning protocol ensures maximum wash efficiency.
Catalog No. 50-255-1978