Learn More
Leica Biosystems Bond DAB Enhancer 30ml
Supplier:  Leica Biosystems AR9432
Encompass
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND DAB Enhancer is a ready to use solution that intensifies the 3,3’-diaminobenzidine (DAB) staining of formalin-fixed, paraffin-embedded tissue on the BOND automated system.DAB is a chromogen used for IHC/ISH staining. In the presence of peroxidase enzyme, DAB produces a brown precipitate, at the epitope site, that is insoluble in alcohol. The BOND DAB Enhancer precipitates as a copper salt at the reaction site producing a darker brown staining deposit.For in vitro diagnostic use.
Catalog No. 50-255-2367