Learn More
Agilent Technologies Bond Elut C18 cartridge, 500 mg, 3 mL, 120 μm, 50 per Pack
Catalog No. NC2945416
Encompass
The most hydrophobic, bonded silica sorbent in the Bond Elut range. Regarded as having the broadest spectrum of retention among bonded silica sorbents. Extremely retentive for nonpolar compounds.
Catalog No. NC2945416 Supplier Agilent Technologies Supplier No. 14102028
The most hydrophobic, bonded silica sorbent in the Bond Elut range. Regarded as having the broadest spectrum of retention among bonded silica sorbents. Extremely retentive for nonpolar compounds.