Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent Technologies Bond Elut SAX, 96 Square-Well Plate, 100 mg, 1/Pk.

Catalog No. NC3804050
Encompass

Agilent Bond Elut SAX is a strong anion exchange SPE plate designed for the extraction of nonpolar and acidic analytes. These 96 square-well plates utilize high-quality silica-based sorbent with a 40 μm particle size for consistent performance in analytical chemistry applications.

  • Phase: SAX (strong anion exchange)
  • Format: 96 square-well plate
  • Sorbent mass: 100 mg per well
  • Well volume: 2 mL
  • Frit material: Polyethylene (20 μm pore size)
  • Plate material: Polypropylene (full skirt)
  • Performance: Retains compounds that elute from weak anion exchange sorbents
  • Flexibility: Selectivity can be user-modified for increased flexibility
  • Typical matrices: Aqueous samples, biological fluids, and buffered organics

Catalog No. NC3804050 Supplier Agilent Technologies Supplier No. A396081C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Agilent Bond Elut SAX is a strong anion exchange SPE plate designed for the extraction of nonpolar and acidic analytes. These 96 square-well plates utilize high-quality silica-based sorbent with a 40 μm particle size for consistent performance in analytical chemistry applications.

  • Phase: SAX (strong anion exchange)
  • Format: 96 square-well plate
  • Sorbent mass: 100 mg per well
  • Well volume: 2 mL
  • Frit material: Polyethylene (20 μm pore size)
  • Plate material: Polypropylene (full skirt)
  • Performance: Retains compounds that elute from weak anion exchange sorbents
  • Flexibility: Selectivity can be user-modified for increased flexibility
  • Typical matrices: Aqueous samples, biological fluids, and buffered organics

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.