Learn More
Leica Biosystems Bond Enzyme Pretreatment Kit
Catalog No. NC1628062
Encompass_Preferred
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Enzyme Pretreatment Kit consists of an enzyme concentrate and an enzyme diluent, which require mixing. The enzyme is diluted before use in the BOND Open Containers supplied. The diluted enzyme solution is used for enzymatic digestion on the automated BOND system.
Catalog No. NC1628062 Supplier Leica Biosystems Supplier No. AR9551
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Enzyme Pretreatment Kit consists of an enzyme concentrate and an enzyme diluent, which require mixing. The enzyme is diluted before use in the BOND Open Containers supplied. The diluted enzyme solution is used for enzymatic digestion on the automated BOND system.