Learn More
Leica Biosystems BOND EPITOPE RETRIEVAL SOLUTIO
Catalog No. NC0235529
Encompass_Preferred
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Epitope Retrieval Solution 1 is a ready to use, citrate based pH 6 epitope retrieval solution for the heat-induced epitope retrieval (HIER) of formalin-fixed, paraffin-embedded tissue on the BOND automated system.The use of a HIER pre-treatment on formalin-fixed, paraffin-embedded tissue on the BOND automated system, restores epitopes that have been modified by formalin fixation, allowing accessibility of the primary antibody to the epitope.For in vitro diagnostic use.
Catalog No. NC0235529 Supplier Leica Biosystems Supplier No. AR9961
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Epitope Retrieval Solution 1 is a ready to use, citrate based pH 6 epitope retrieval solution for the heat-induced epitope retrieval (HIER) of formalin-fixed, paraffin-embedded tissue on the BOND automated system.The use of a HIER pre-treatment on formalin-fixed, paraffin-embedded tissue on the BOND automated system, restores epitopes that have been modified by formalin fixation, allowing accessibility of the primary antibody to the epitope.For in vitro diagnostic use.