Learn More
Leica Biosystems BOND FISH Kit
Supplier:  Leica Biosystems DS9374
Encompass
BOND FISH KitThe BOND FISH Kit enables the user to perform fluorescence in situ hybridisation (FISH) on the automated BOND system.It is intended for use with nucleic acid probes on formalin-fixed, paraffin embedded (FFPE) tissue. The kit consists of a formamide mixture which reduces non-specific hybridisation of nucleic acid probes.Total volume = 18 mL, sufficient for 60 tests.
Catalog No. 50-255-2273