Learn More
Leica Biosystems BOND INTENSE R DETECT SYSTEM
Catalog No. NC2459083
Encompass
  • Peroxidase detection system optimized for automated use.
  • Research use only application focus.
  • Compatible with automated BOND systems.
  • Detection chemistry utilizes peroxidase (HRP).
  • Chromogen produces brown staining (DAB).
  • Includes peroxide block and hematoxylin counterstain.
  • Requires user-supplied biotinylated secondary antibody.
  • Offers strong staining intensity.
  • Supports any primary antibody species.
Catalog No. NC2459083 Supplier Leica Biosystems Supplier No. DS9263
  • Peroxidase detection system optimized for automated use.
  • Research use only application focus.
  • Compatible with automated BOND systems.
  • Detection chemistry utilizes peroxidase (HRP).
  • Chromogen produces brown staining (DAB).
  • Includes peroxide block and hematoxylin counterstain.
  • Requires user-supplied biotinylated secondary antibody.
  • Offers strong staining intensity.
  • Supports any primary antibody species.