Learn More
Leica Biosystems BOND OPEN CONTAINERS 7ML
Catalog No. NC0498827
Encompass
BOND Open 7 mL Containers allow the use of reagents from any source on the BOND system.Each container can be refilled until a total of 4 mL has been dispensed from it.They are ideal for reagents that are consumed intermittently and have a short shelf life.
Catalog No. NC0498827 Supplier Leica Biosystems Supplier No. OP79193
BOND Open 7 mL Containers allow the use of reagents from any source on the BOND system.Each container can be refilled until a total of 4 mL has been dispensed from it.They are ideal for reagents that are consumed intermittently and have a short shelf life.