Learn More
Leica Biosystems BOND OPEN CONTAINERS 30ML
Catalog No. NC0829813
Encompass
BOND Open 3 mL Containers allow the use of reagents from any source on the BOND system.Each container holds 3 mL and can be refilled until a total of 4 mL has been dispensed from it.They are ideal for high throughput reagents that are consumed on a daily basis and their use can minimize reagent preparation time.
Catalog No. NC0829813 Supplier Leica Biosystems Supplier No. OP309700
BOND Open 3 mL Containers allow the use of reagents from any source on the BOND system.Each container holds 3 mL and can be refilled until a total of 4 mL has been dispensed from it.They are ideal for high throughput reagents that are consumed on a daily basis and their use can minimize reagent preparation time.