Learn More
Leica Biosystems BOND PLUS SLIDES 1440/BOX
Catalog No. NC1823288
Encompass_Preferred
BOND Plus Slides are positively charged glass microscopic slides designed for use on the Bond system. They include defined margins to enable the accurate placement of tissue for staining in the 100 µL and the 150 µL dispense modes, which helps in maintaining the integrity of staining quality.
Catalog No. NC1823288 Supplier Leica Biosystems Supplier No. S21.2113.A
BOND Plus Slides are positively charged glass microscopic slides designed for use on the Bond system. They include defined margins to enable the accurate placement of tissue for staining in the 100 µL and the 150 µL dispense modes, which helps in maintaining the integrity of staining quality.