Learn More
Leica Biosystems BOND PRIMARY ANTIBODY DILUENT
Catalog No. NC0235532
Encompass
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Primary Antibody Diluent is a ready-to-use reagent for the dilution of individual primary antibody concentrates for use on the BOND.It is not to be used for the reconstitution of lyophilized antibodies.For in vitro diagnostic use.
Catalog No. NC0235532 Supplier Leica Biosystems Supplier No. AR9352
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Primary Antibody Diluent is a ready-to-use reagent for the dilution of individual primary antibody concentrates for use on the BOND.It is not to be used for the reconstitution of lyophilized antibodies.For in vitro diagnostic use.