Learn More
Leica Biosystems BOND PRINTER RIBBON AND LABELS
Catalog No. NC2187751
Encompass
The BOND Slide Label and Print Ribbon Kit produces high-quality solvent-resistant slide labels for the use on the BOND system. This assists in preserving the integrity of slide and patient record identification on BOND slides.The BOND Universal Slide Labels adhere to slides for easy and secure identification.Reagent CompatibilityExtended soaking in, or exposure to, chemicals can reduce the effectiveness of the slide label and may result in label damage, lifting, or loss of print integrity. It is recommended that labels are not submerged in fluid for an extended period and label usage is validated by the laboratory.
This item is not returnable. View return policy
Catalog No. NC2187751 Supplier Leica Biosystems Supplier No. S21.4604
The BOND Slide Label and Print Ribbon Kit produces high-quality solvent-resistant slide labels for the use on the BOND system. This assists in preserving the integrity of slide and patient record identification on BOND slides.The BOND Universal Slide Labels adhere to slides for easy and secure identification.Reagent CompatibilityExtended soaking in, or exposure to, chemicals can reduce the effectiveness of the slide label and may result in label damage, lifting, or loss of print integrity. It is recommended that labels are not submerged in fluid for an extended period and label usage is validated by the laboratory.