Learn More
Leica Biosystems Bond Polymer Refine Red Detection
Catalog No. NC0348028
Encompass
BOND Polymer Refine Red Detection is an IVD labeled red detection system for the automated BOND system.BOND Polymer Refine Red Detection is biotin-free, utilizing alkaline phosphatase (AP)-linked compact polymers to provide enhanced tissue penetration and unsurpassed reagent sensitivity.It contains post primary, polymer reagent, Fast Red chromogen, and hematoxylin counterstain and is supplied in a convenient, ready-to-use format.
Catalog No. NC0348028 Supplier Leica Biosystems Supplier No. DS9390
BOND Polymer Refine Red Detection is an IVD labeled red detection system for the automated BOND system.BOND Polymer Refine Red Detection is biotin-free, utilizing alkaline phosphatase (AP)-linked compact polymers to provide enhanced tissue penetration and unsurpassed reagent sensitivity.It contains post primary, polymer reagent, Fast Red chromogen, and hematoxylin counterstain and is supplied in a convenient, ready-to-use format.