Learn More
Leica Biosystems Bond Slide Labeler -
Supplier:  Leica Biosystems S21.1913
Encompass
The BOND Slide Labeler Cleaning Pen is used to clean the print head on the BOND Slide Labeler.Regular cleaning helps ensure labels are printed clearly and correctly.
Catalog No. 50-255-1975