Learn More
Leica Biosystems BOND TITRATION CONTAINER 50/P
Catalog No. NC0348036
Encompass
BOND Titration Container Inserts are tubes that fit directly into the BOND Titration Containers.They enable safer use of up to 4 mL of reagent per titration container.
Catalog No. NC0348036 Supplier Leica Biosystems Supplier No. OPT9719
BOND Titration Container Inserts are tubes that fit directly into the BOND Titration Containers.They enable safer use of up to 4 mL of reagent per titration container.