Learn More
Leica Biosystems BOND UNIVERSAL COVERTILES
Catalog No. NC1206859
Encompass
Antigen BackgroundThe BOND Universal Covertile is a patented technology that facilitates gentle, even reagent flow over tissue. It prevents reagent evaporation and minimizes waste generation. The Covertile is re-usable and can also be recycled once its staining life is over.
Catalog No. NC1206859 Supplier Leica Biosystems Supplier No. S21.4611
Antigen BackgroundThe BOND Universal Covertile is a patented technology that facilitates gentle, even reagent flow over tissue. It prevents reagent evaporation and minimizes waste generation. The Covertile is re-usable and can also be recycled once its staining life is over.