Learn More
Waters Corp Prep Column, Bondapak C18, 15-20 μm, 19 x 300 mm
Catalog No. 50785219 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Bondapak C18 is available in two different particle sizes, 15-20 μm and 37-55 μm. This allows for easy method transfer from μBondapak through preparative scale-up. It provides an optimal balance for developing new purification protocols.
  • Available in two particle sizes (15-20 μm and 37-55 μm) for method transfer to preparative scale.
  • Provides the best balance for developing new purification protocols.
Catalog No. 50-785-219 Supplier Waters Corp Supplier No. WAT025834
Bondapak C18 is available in two different particle sizes, 15-20 μm and 37-55 μm. This allows for easy method transfer from μBondapak through preparative scale-up. It provides an optimal balance for developing new purification protocols.
  • Available in two particle sizes (15-20 μm and 37-55 μm) for method transfer to preparative scale.
  • Provides the best balance for developing new purification protocols.