Learn More
Waters Corp Waters Cartridge, C18, 15-20 μm, 47 mm x 300 mm
Catalog No. 50785976 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
PrepPAK columns are plastic compression cartridge segments for liquid chromatography systems. This general-purpose, silica-based, reversed-phase C18 column utilizes 15-20μm particle technology, balancing resolution, throughput, and cost for new purification protocols.
  • Designed as plastic compression cartridge segments for use with PrepPAK compression modules (holder sold separately).
  • Silica-based, reversed-phase C18 column with 15-20μm particle technology.
  • Available in 15-20μm and 37-55μm particle sizes for easy method transfer to preparative scale-up.
  • Manufactured under strict cGMP and ISO 9002 conditions, ensuring high quality, consistency, and reliable analysis results.
  • Provides excellent column efficiency, peak symmetry, column lifetime, and column-to-column reproducibility.
Catalog No. 50-785-976 Supplier Waters Corp Supplier No. WAT091784
PrepPAK columns are plastic compression cartridge segments for liquid chromatography systems. This general-purpose, silica-based, reversed-phase C18 column utilizes 15-20μm particle technology, balancing resolution, throughput, and cost for new purification protocols.
  • Designed as plastic compression cartridge segments for use with PrepPAK compression modules (holder sold separately).
  • Silica-based, reversed-phase C18 column with 15-20μm particle technology.
  • Available in 15-20μm and 37-55μm particle sizes for easy method transfer to preparative scale-up.
  • Manufactured under strict cGMP and ISO 9002 conditions, ensuring high quality, consistency, and reliable analysis results.
  • Provides excellent column efficiency, peak symmetry, column lifetime, and column-to-column reproducibility.