Learn More
Waters Corp Bondapak C18 Radial-Pak Column, 125Å, 15 - 20 µm, 8 mm X 100 mm, 1/pk
Catalog No. 50785221 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Bondapak C18 is available in two different particle sizes, 15-20µm and 37-55µm, providing easy method transfer from µBondapak through preparative scale up. If you are developing a new purification protocol, Bondapak provides the best balance between resolution, throughput and cost.
Catalog No. 50-785-221 Supplier Waters Corp Supplier No. WAT025841
Bondapak C18 is available in two different particle sizes, 15-20µm and 37-55µm, providing easy method transfer from µBondapak through preparative scale up. If you are developing a new purification protocol, Bondapak provides the best balance between resolution, throughput and cost.