Learn More
Waters Corp Waters Bondapak Column, C18, 300Å, 15–20 μm
Catalog No. 50785475 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Bondapak C18 is available in two different particle sizes, 15–20μm and 37–55μm, offering easy method transfer from μBondapak through preparative scale up. This product provides an optimal balance between resolution, throughput, and cost for those developing new purification protocols.
  • Available in two particle sizes: 15–20μm and 37–55μm.
  • Facilitates easy method transfer from μBondapak to preparative scale up.
  • Offers the best balance of resolution, throughput, and cost.
Catalog No. 50-785-475 Supplier Waters Corp Supplier No. WAT038575
Bondapak C18 is available in two different particle sizes, 15–20μm and 37–55μm, offering easy method transfer from μBondapak through preparative scale up. This product provides an optimal balance between resolution, throughput, and cost for those developing new purification protocols.
  • Available in two particle sizes: 15–20μm and 37–55μm.
  • Facilitates easy method transfer from μBondapak to preparative scale up.
  • Offers the best balance of resolution, throughput, and cost.