Learn More
Waters Corp Waters C18 Column, 15 to 20 μm, 3.9 mm X 300 mm
Catalog No. 50785246 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Bondapak C18 is available in two different particle sizes: 15 to 20 μm and 37 to 55 μm, allowing for easy method transfer from μBondapak through preparative scale up. It provides the best balance between resolution, throughput, and cost for developing a new purification protocol. Waters is the sole manufacturer of authentic analytical columns using Bondapak C18 packing material, emphasizing stringent quality control and nearly fifty years of proven year-to-year repeatability.
  • Provides the best balance between resolution, throughput, and cost for developing a new purification protocol.
  • Offers the finest blend of resolution, throughput, and cost for establishing a novel purification technique.
  • Manufactured with authentic C18 packing material.
  • Packing materials have proved year-to-year repeatability for nearly fifty years.
Catalog No. 50-785-246 Supplier Waters Corp Supplier No. WAT025875
Bondapak C18 is available in two different particle sizes: 15 to 20 μm and 37 to 55 μm, allowing for easy method transfer from μBondapak through preparative scale up. It provides the best balance between resolution, throughput, and cost for developing a new purification protocol. Waters is the sole manufacturer of authentic analytical columns using Bondapak C18 packing material, emphasizing stringent quality control and nearly fifty years of proven year-to-year repeatability.
  • Provides the best balance between resolution, throughput, and cost for developing a new purification protocol.
  • Offers the finest blend of resolution, throughput, and cost for establishing a novel purification technique.
  • Manufactured with authentic C18 packing material.
  • Packing materials have proved year-to-year repeatability for nearly fifty years.