Learn More
Chemglass Life Sciences Bonded Closure, Black, 8-425, PTFE/Silicone with Slit
SDP
Catalog No. 50231103
Encompass_Preferred
Bonded Closure, Black, 8-425, PTFE/Silicone with Slit. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Pre-slit design.
Catalog No. 50-231-103 Supplier Chemglass Life Sciences Supplier No. CV38420008
Bonded Closure, Black, 8-425, PTFE/Silicone with Slit. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Pre-slit design.