Learn More
Chemglass Life Sciences Standard Bonded Closure, 9mm, Blue, PTFE/Silicone Septa
SDP
Catalog No. 50231112
Encompass_Preferred
Standard Bonded Closure, 9mm, Blue, PTFE/Silicone Septa. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit design.
Catalog No. 50-231-112 Supplier Chemglass Life Sciences Supplier No. CV3845B009
Standard Bonded Closure, 9mm, Blue, PTFE/Silicone Septa. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Non-slit design.