Learn More
Chemglass Life Sciences Standard Bonded Closure, 9mm, Black, PTFE/Silicone Septa with Slit
SDP
Catalog No. 50231113
Encompass_Preferred
Standard Bonded Closure, 9mm, Black, PTFE/Silicone Septa with Slit. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Pre-slit design.
Catalog No. 50-231-113 Supplier Chemglass Life Sciences Supplier No. CV3845BS09
Standard Bonded Closure, 9mm, Black, PTFE/Silicone Septa with Slit. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout. Pre-slit design.