Learn More
Chemglass Life Sciences Bonded Closure, 9mm, Gray, PTFE/Silicone Septa with Slit, for MASS SPEC ILT LC-CLEAN
SDP
Catalog No. 501532578
Encompass_Preferred
Bonded Closure, 9mm, Gray, PTFE/Silicone Septa with Slit, for MASS SPEC ILT LC-CLEAN. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.
Catalog No. 50-153-2578 Supplier Chemglass Life Sciences Supplier No. CV38430009
Bonded Closure, 9mm, Gray, PTFE/Silicone Septa with Slit, for MASS SPEC ILT LC-CLEAN. Bonded closures feature clear PTFE/Silicone liner bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.