Learn More
Chemglass Life Sciences Bonded Closure, White, 10-425, PTFE/Silicone
SDP
Catalog No. 501532576
Encompass_Preferred
Bonded Closure, White, 10-425, PTFE/Silicone. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.
Catalog No. 50-153-2576 Supplier Chemglass Life Sciences Supplier No. CV3841W010
Bonded Closure, White, 10-425, PTFE/Silicone. Bonded closures feature clear PTFE/Silicone bonded directly to the closure without the use of glue or adhesives. These bonded closures provide a cost effective, integrated, one-piece system that eliminates costly line fallout.