Learn More
Leica Biosystems BONDTM WASH SOLUTION 10X
Catalog No. NC0221077
Encompass
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Wash Solution 1X Concentrate is a concentrated buffer solution, requiring initial dilution. The diluted solution is for washing sections of formalin-fixed, paraffin-embedded tissue during staining on the BOND automated system.The BOND automated system requires the use of a specific wash buffer to remove unbound material at the end of each incubation step. This buffer is prepared by dilution, using BOND Wash Solution 1X Concentrate. The appropriate bulk container is then filled and placed within the BOND Processing Module.For in vitro diagnostic use.
Catalog No. NC0221077 Supplier Leica Biosystems Supplier No. AR9590
BOND ancillaries have been designed for Leica Biosystems fully-automated BOND systems: BOND-MAX and BOND-III systems. Used in the development and validation of BOND ready-to-use primary antibodies and ISH probes - choose Leica Biosystems reagents for the assurance of a quality result in your laboratory.BOND Wash Solution 1X Concentrate is a concentrated buffer solution, requiring initial dilution. The diluted solution is for washing sections of formalin-fixed, paraffin-embedded tissue during staining on the BOND automated system.The BOND automated system requires the use of a specific wash buffer to remove unbound material at the end of each incubation step. This buffer is prepared by dilution, using BOND Wash Solution 1X Concentrate. The appropriate bulk container is then filled and placed within the BOND Processing Module.For in vitro diagnostic use.