Learn More
Medchemexpress LLC Bonducellpin D | 197781-85-4 | 1 MG
SDP
Catalog No. 5000421741
Encompass_Preferred
Bonducellpin D is a furanoditerpenoid lactone isolated from Caesalpinia minax. It demonstrates broad-spectrum inhibition against SARS-CoV Mpro and MERS-CoV Mpro, with Ki values of 467.11 nM and 284.86 nM, respectively. This compound also exhibits moderate anti-cancer activity in vitro.
  • Isolated from Caesalpinia minax
  • Demonstrates broad-spectrum inhibition potential against SARS-CoV Mpro
  • Inhibits MERS-CoV Mpro
  • Exhibits moderate anti-cancer activity in vitro
Catalog No. 50-004-21741 Supplier Medchemexpress LLC Supplier No. HYN29491MG
Bonducellpin D is a furanoditerpenoid lactone isolated from Caesalpinia minax. It demonstrates broad-spectrum inhibition against SARS-CoV Mpro and MERS-CoV Mpro, with Ki values of 467.11 nM and 284.86 nM, respectively. This compound also exhibits moderate anti-cancer activity in vitro.
  • Isolated from Caesalpinia minax
  • Demonstrates broad-spectrum inhibition potential against SARS-CoV Mpro
  • Inhibits MERS-CoV Mpro
  • Exhibits moderate anti-cancer activity in vitro