Learn More
Med Vet International Bone Chisel, 6mm, J0227A
Catalog No. 5000584380
Encompass_Preferred
Striking button for easier use with Gerzog mallet. German-made. Stainless steel. 17cm L (6 1/2").
Catalog No. 50-005-84380 Supplier Med Vet International Supplier No. JOR227A
Striking button for easier use with Gerzog mallet. German-made. Stainless steel. 17cm L (6 1/2").