Learn More
Med Vet International Bone Clip Plate, 3.5 mm, Each
Catalog No. 5000582817
Encompass_Preferred
The storage of plates in an orderly fashion is desirable. This inexpensive clip allows plate storage with quick access while making proper plate selection easier.Holds up to eight platesID tag identifies each plate size
Catalog No. 50-005-82817 Supplier Med Vet International Supplier No. JOR430PC
The storage of plates in an orderly fashion is desirable. This inexpensive clip allows plate storage with quick access while making proper plate selection easier.Holds up to eight platesID tag identifies each plate size