Learn More
World Precision Instrument Bone Cutter, 10.5cm, Straight, Strong/Serrated Tips
SDP
Catalog No. 502539529
Encompass
WPI Bone Cutters have short, thick scissor-style blades, which are designed to efficiently cut through bone tissue. Unlike typical scissor blades, these forceps have a more robust, heavier structure to enable effective cutting and shaping of bone during surgical procedures. The ring handles offer a comfortable grip for the surgeon and provide precise control during bone-cutting procedures. These are 10.5 cm (4.13”) long and are available with curved or straight blades, and smooth or serrated tips. The are made of surgical grade stainless steel, which is suitable for chemical sterilization or autoclaving. Clean your surgical instruments promptly after use to ensure a long life.
Catalog No. 50-253-9529 Supplier World Precision Instrument Supplier No. 503253
WPI Bone Cutters have short, thick scissor-style blades, which are designed to efficiently cut through bone tissue. Unlike typical scissor blades, these forceps have a more robust, heavier structure to enable effective cutting and shaping of bone during surgical procedures. The ring handles offer a comfortable grip for the surgeon and provide precise control during bone-cutting procedures. These are 10.5 cm (4.13”) long and are available with curved or straight blades, and smooth or serrated tips. The are made of surgical grade stainless steel, which is suitable for chemical sterilization or autoclaving. Clean your surgical instruments promptly after use to ensure a long life.