Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw
SDP

Catalog No. 22444344 Shop All Mopec Products
Change view
Click to view available options
For Use With (Equipment):
Stryker Saw
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22-444-344 Stryker Saw
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-444-344 Supplier Mopec™ Supplier No. BD080
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mopec Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw features Hepa filtration to minimize exposure to particulate matter. Attaches directly to the autopsy saw for safety and efficiency.

  • Hepa Vacuum includes Hepa Filter, Vacuum Hose (102in.) and Y Bracket
  • One Year Warranty
  • 115V, 60Hz with US plug

Warranty and Services

120V input, 0-140V, 15.00A

Specifications

Frequency 60 Hz
For Use With (Equipment) Stryker Saw
Voltage 115 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.