Learn More
Med Vet International Bone Fragment Forceps 6" Stainless Steel, J0212
Catalog No. 5000584412
Encompass_Preferred
Bone Fragment (Reduction) Forceps. Fragment forceps are used for temporary accurate reduction of fragments while implants are being applied.Fragment Forceps. Standard. 6" (15cm).
Catalog No. 50-005-84412 Supplier Med Vet International Supplier No. JOR212
Bone Fragment (Reduction) Forceps. Fragment forceps are used for temporary accurate reduction of fragments while implants are being applied.Fragment Forceps. Standard. 6" (15cm).