Learn More
CPC Scientific H-Phe-Tyr-Gly-Pro-Val-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific OSTP004B
Encompass
SEQUENCE: H-Phe-Tyr-Gly-Pro-Val-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: FYGPVMOLECULAR FORMULA: C30H39N5O7MOLECULAR WEIGHT: 581.66STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [85679-70-5]SYNONYMS: Osteocalcin (45-49) (human)RESEARCH AREA: OsteoporosisREFERENCES:SKU(s): OSTP-004A, OSTP-004B
Catalog No. 50-211-0006