Learn More
Med Vet International Bone Holding Forcep, 6in, J0213, Each
Catalog No. 5000585541
Encompass_Preferred
Stainless steel. 15 cm. 6".For small dogs and catsWide jaws offer maximum compressionBone screws and IM pins can be inserted through the center
Catalog No. 50-005-85541 Supplier Med Vet International Supplier No. JOR213
Stainless steel. 15 cm. 6".For small dogs and catsWide jaws offer maximum compressionBone screws and IM pins can be inserted through the center