Learn More
World Precision Instrument Bone Mallet, Post Mortem, Hook Handle, 20.3 cm
SDP
Catalog No. 502538377
Encompass
The Bone Mallet is a special hammer for forensic and post-mortem work. Often used to tap bone chisels to apply the necessary force. The hook at the end of the handle can be used to separate the calvarium from the bottom section of the skull when accessing the brain. This hammer is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8377 Supplier World Precision Instrument Supplier No. WP0059
The Bone Mallet is a special hammer for forensic and post-mortem work. Often used to tap bone chisels to apply the necessary force. The hook at the end of the handle can be used to separate the calvarium from the bottom section of the skull when accessing the brain. This hammer is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.