Learn More
Sigma Aldrich Fine Chemicals Biosciences Bone meal NISTR SRMR 1486
Catalog No. 502908062
Encompass
This Standard Reference Material (SRM) is intended primarily for use in evaluating analytical methods used for the determination of selected major minor and trace elements in bone and in material of a similar matrix. It consists of steamed bone meal that was sieved and blended to a high degree of homogeneity. A unit of SRM 1486 consists ofapproximately 50 g of bone meal. For more information please refer to the SDS and COA.SRM 1486 cert SRM 1486 SDS
Catalog No. 50-290-8062 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NIST1486
This Standard Reference Material (SRM) is intended primarily for use in evaluating analytical methods used for the determination of selected major minor and trace elements in bone and in material of a similar matrix. It consists of steamed bone meal that was sieved and blended to a high degree of homogeneity. A unit of SRM 1486 consists ofapproximately 50 g of bone meal. For more information please refer to the SDS and COA.SRM 1486 cert SRM 1486 SDS