Learn More
Cusabio Technology LLC bone morphogenetic protein 15
Catalog No. 5000050009
Encompass_Preferred
This kit detects BMP15 in human serum plasma and cell culture supernates with a sensitivity of 3 9 pg/mL and a quantitative detection range of 15 6 1000 pg/mL Utilizing a sandwich ELISA methodology it requires a sample volume of 50 100 L and features a streamlined protocol with incubation times between 1 5 hours The assay is measured at 450 nm providing reliable results for research or diagnostic applications requiring precise measurement of BMP15/GDF-9B levels in biological samples Its robust design ensures high specificity and reproducibility making it suitable for studies focused on reproductive biology ovarian function or related clinical investigations
Catalog No. 50-000-50009 Supplier Cusabio Technology LLC Supplier No. CSBE17529H
This kit detects BMP15 in human serum plasma and cell culture supernates with a sensitivity of 3 9 pg/mL and a quantitative detection range of 15 6 1000 pg/mL Utilizing a sandwich ELISA methodology it requires a sample volume of 50 100 L and features a streamlined protocol with incubation times between 1 5 hours The assay is measured at 450 nm providing reliable results for research or diagnostic applications requiring precise measurement of BMP15/GDF-9B levels in biological samples Its robust design ensures high specificity and reproducibility making it suitable for studies focused on reproductive biology ovarian function or related clinical investigations