Learn More
Cusabio Technology LLC bone morphogenetic protein 7
Catalog No. 5000049511
Encompass_Preferred
This kit detects BMP7 in human serum plasma and cell culture supernates with a quantitative Sandwich ELISA method It offers a broad detection range of 15 6 pg/mL to 1000 pg/mL and demonstrates high sensitivity with a lower limit of detection at 3 9 pg/mL The assay requires a short incubation period of 1-5 hours and utilizes sample volumes between 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable detection of BMP7 concentrations across biological matrices Designed for research applications this kit provides precise quantification of BMP7 levels to support studies in bone metabolism renal development and tissue regeneration pathways
Catalog No. 50-000-49511 Supplier Cusabio Technology LLC Supplier No. CSBE08122H
This kit detects BMP7 in human serum plasma and cell culture supernates with a quantitative Sandwich ELISA method It offers a broad detection range of 15 6 pg/mL to 1000 pg/mL and demonstrates high sensitivity with a lower limit of detection at 3 9 pg/mL The assay requires a short incubation period of 1-5 hours and utilizes sample volumes between 50-100 L per test Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable detection of BMP7 concentrations across biological matrices Designed for research applications this kit provides precise quantification of BMP7 levels to support studies in bone metabolism renal development and tissue regeneration pathways