Learn More
Cusabio Technology LLC bone morphogenetic protein 7
Catalog No. 5000050341
Encompass_Preferred
This kit detects BMP7 in mouse serum plasma cell culture supernates and tissue homogenates offering a quantitative measurement range of 0 625 pg/mL to 40 pg/mL with a sensitivity of 0 156 pg/mL Utilizing a sandwich ELISA methodology it provides reliable detection within 1-5 hours using 50-100 L of sample per test The assay is read at an optical density of 450 nm ensuring precise and reproducible results for research applications requiring accurate BMP7 level analysis across diverse biological matrices
Catalog No. 50-000-50341 Supplier Cusabio Technology LLC Supplier No. CSBE08125M
This kit detects BMP7 in mouse serum plasma cell culture supernates and tissue homogenates offering a quantitative measurement range of 0 625 pg/mL to 40 pg/mL with a sensitivity of 0 156 pg/mL Utilizing a sandwich ELISA methodology it provides reliable detection within 1-5 hours using 50-100 L of sample per test The assay is read at an optical density of 450 nm ensuring precise and reproducible results for research applications requiring accurate BMP7 level analysis across diverse biological matrices