Learn More
Med Vet International Bone Plate Benders for Small Bender, 1.5mm/2.0mm. Usedfor small 1.5/2.0mm plates. 140mm L, J0853, Each
Catalog No. 5000584987
Encompass_Preferred
Proper contouring of bone plates is a necessity for proper reduction of fractures. Used in pairs, bone plate benders are the most cost-effective method of plate contouring. Stainless steel.
Catalog No. 50-005-84987 Supplier Med Vet International Supplier No. JOR853
Proper contouring of bone plates is a necessity for proper reduction of fractures. Used in pairs, bone plate benders are the most cost-effective method of plate contouring. Stainless steel.